missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRASP65 Recombinant Rabbit Monoclonal Antibody (23GB5580), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553902
This item is not returnable.
View return policy
Description
Grasp65 is a structural protein of the golgi apparatus. It has been implicated in the stacking of golgi cisternae. Grasp65 is shown to have the properties to bind surfaces together in a mitotically regulated matter.
Specifications
| GRASP65 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| GORASP1 | |
| 5430411C10Rik; Golgi peripheral membrane protein p65; Golgi phosphoprotein 5; Golgi reassembly and stacking protein 1; golgi reassembly stacking protein 1; golgi reassembly stacking protein 1, 65kDa; Golgi reassembly-stacking protein 1; Golgi reassembly-stacking protein of 65 kDa; GOLPH5; Gorasp1; GRASP65; P65 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 64689 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5580 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BQQ3 | |
| GORASP1 | |
| A synthesized peptide derived from human GRASP65 (400-440AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction