missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRASP65 Recombinant Rabbit Monoclonal Antibody (23GB5580), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | GRASP65 |
|---|---|
| Clone | 23GB5580 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089083
![]() |
Invitrogen™
MA553902 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087410
|
Invitrogen™
MA553903 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRASP65 Recombinant Monoclonal antibody specifically detects GRASP65 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| GRASP65 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9BQQ3 | |
| 64689 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5580 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 5430411C10Rik; Golgi peripheral membrane protein p65; Golgi phosphoprotein 5; Golgi reassembly and stacking protein 1; golgi reassembly stacking protein 1; golgi reassembly stacking protein 1, 65kDa; Golgi reassembly-stacking protein 1; Golgi reassembly-stacking protein of 65 kDa; GOLPH5; Gorasp1; GRASP65; P65 | |
| A synthesized peptide derived from human GRASP65 (400-440AA). | |
| Primary | |
| -20°C | |
| GORASP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title