missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRASP65/GORASP1 Mouse anti-Human, Mouse, Rat, Clone: 1B2C10, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | GRASP65/GORASP1 |
|---|---|
| Clone | 1B2C10 |
| Concentration | 0.8 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16888725
|
Proteintech
66651-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16878725
|
Proteintech
66651-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Grasp65 is a structural protein of the golgi apparatus. It has been implicated in the stacking of golgi cisternae. Grasp65 is shown to have the properties to bind surfaces together in a mitotically regulated matter.Specifications
| GRASP65/GORASP1 | |
| 0.8 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| FLJ23443, Golgi phosphoprotein 5, GOLPH5, GORASP1, GRASP65, P65 | |
| GORASP1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1B2C10 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Rat, Mouse, Human | |
| O35254, Q91X51, Q9BQQ3 | |
| 56082, 64689, 74498 | |
| GRASP65,GORASP1 Fusion Protein Ag1054 | |
| Primary | |
| -20°C | |
| GORASP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title