missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRASP65/GORASP1 Mouse anti-Human, Mouse, Rat, Clone: 1B2C10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66651-1-IG-20UL
This item is not returnable.
View return policy
Description
Grasp65 is a structural protein of the golgi apparatus. It has been implicated in the stacking of golgi cisternae. Grasp65 is shown to have the properties to bind surfaces together in a mitotically regulated matter.Specifications
| GRASP65/GORASP1 | |
| Monoclonal | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35254, Q91X51, Q9BQQ3 | |
| GORASP1 | |
| GRASP65,GORASP1 Fusion Protein Ag1054 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1B2C10 | |
| Unconjugated | |
| GORASP1 | |
| FLJ23443, Golgi phosphoprotein 5, GOLPH5, GORASP1, GRASP65, P65 | |
| Mouse | |
| Protein G | |
| RUO | |
| 56082, 64689, 74498 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction