missing translation for 'onlineSavingsMsg'
Learn More

Graphite rod, pyrolytic coated, 2mm (0.08in) dia x 152mm (6in) long, Thermo Scientific Chemicals

Product Code. 15446545
Change view
Click to view available options
Quantity:
1 Pc.
5 Pc.
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446545 5 Pc. Pack of 5
15436545 1 Pc. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15446545 Supplier Thermo Scientific Alfa Aesar Supplier No. 043670.KJ
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Graphite rod, is suitable for making: Anodes, Electrodes, Heating elements, Crucibles, Molds & Ingots, Hot press dies, Gas nozzles, Lubrication sticks, Bushings & bearings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Graphite rod
Name Note Pyrolytic coated, 2mm (0.08 in.) dia. x 152mm (6 in.) long
MDL Number MFCD00144065
Quantity 5 Pc.
Solubility Information It is insoluble in water.
Physical Form Rod

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.