missing translation for 'onlineSavingsMsg'
Learn More

Graphite rod, pyrolytic coated, 2mm (0.08in) dia x 152mm (6in) long, Thermo Scientific Chemicals

Product Code. 15436545
Click to view available options
Quantity:
1 Pc.
5 Pc.
Unit Size:
Each
Pack of 5
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15436545

Brand: Thermo Scientific Alfa Aesar 043670.KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Graphite rod, is suitable for making: Anodes, Electrodes, Heating elements, Crucibles, Molds & Ingots, Hot press dies, Gas nozzles, Lubrication sticks, Bushings & bearings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Graphite rod
Name Note Pyrolytic coated, 2mm (0.08 in.) dia. x 152mm (6 in.) long
MDL Number MFCD00144065
Quantity 1 Pc.
Solubility Information It is insoluble in water.
Physical Form Rod

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.