missing translation for 'onlineSavingsMsg'
Learn More

Graphite rod, 3.05mm (0.12in) dia x 305mm (12in) long, 99.9995% (metals basis), Thermo Scientific Chemicals

Product Code. 11430341
Change view
Click to view available options
Quantity:
100 Pc.
25 Pc.
Unit Size:
Pack of 100
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430341 25 Pc. Pack of 25
11440341 100 Pc. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11430341 Supplier Thermo Scientific Alfa Aesar Supplier No. 040765.KM
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Graphite rod is used in computer brushes, anodes, arc lamps, carbon coating, cathode ray tubes, and moderator in nuclear piles. It is used for high-temperature crucibles, as a lubricant and in lead pencils. Graphite rod coated with Th(IV), plays an useful application for ion selective electrode based on thorium oxinate complex. It is also essential for the preparation of lead(II)-selective PVC-coated graphite rod electrode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Commutar brushes, anodes, arc lamps, carbon coating, cathode ray tubes, moderator in nuclear piles
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Graphite rod
Name Note 3.05mm (0.12 in.) dia. x 305mm (12 in.) long
Purity Grade Notes AGKSP grade, ultra 'F' purity
Assay Percent Notes (metals basis)
MDL Number MFCD00144065
Quantity 25 Pc.
Solubility Information Insoluble in water. Soluble in molten nickel.
Percent Purity 99.9995%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.