Learn More
Graphite rod, 3.05mm (0.12in) dia x 305mm (12in) long, 99.9995% (metals basis), Thermo Scientific Chemicals
Description
Graphite rod is used in computer brushes, anodes, arc lamps, carbon coating, cathode ray tubes, and moderator in nuclear piles. It is used for high-temperature crucibles, as a lubricant and in lead pencils. Graphite rod coated with Th(IV), plays an useful application for ion selective electrode based on thorium oxinate complex. It is also essential for the preparation of lead(II)-selective PVC-coated graphite rod electrode.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Commutar brushes, anodes, arc lamps, carbon coating, cathode ray tubes, moderator in nuclear piles
Specifications
Specifications
| Chemical Name or Material | Graphite rod |
| Name Note | 3.05mm (0.12 in.) dia. x 305mm (12 in.) long |
| Purity Grade Notes | AGKSP grade, ultra 'F' purity |
| Assay Percent Notes | (metals basis) |
| MDL Number | MFCD00144065 |
| Quantity | 100 Pc. |
| Solubility Information | Insoluble in water. Soluble in molten nickel. |
| Percent Purity | 99.9995% |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.