missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Graphite powder, natural, universal grade, -200 mesh, 99.9995% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
28 g
113 g
227 g
454 g
Description
Used in aircraft manufacturing industries, defense material, automotive industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Black |
Mesh Size | −200 Mesh |
Odor | Odorless |
Quantity | 113 g |
UN Number | UN1325 |
Solubility Information | Insoluble. |
Percent Purity | 99.9995% |
Grade | UCP-1-U grade, ultra 'F' purity |
Physical Form | Powder |
Assay | (metals basis) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'