missing translation for 'onlineSavingsMsg'
Learn More

Graphite powder, natural, microcrystal grade, APS 2-15 micron, 99.9995% (metals basis), Thermo Scientific Chemicals

Product Code. p-7049573
Click to view available options
Quantity:
28 g
113 g
227 g
454 g
Unit Size:
113g
227g
28g
454g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11367068

Brand: Thermo Scientific Alfa Aesar 014736.TA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

 

Graphite powder is used to make crucibles, lead pencils, electrodes, lubricants, stove polish, brake linings, batteries, glass manufacture, conductive coatings, and for foundry facings. It is used in the synthesis of single sheet graphene nanoflakes and graphite based magnetorheological elastomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-42-5
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Color Black
Odor Odorless
Quantity 28 g
UN Number UN1325
Solubility Information Insoluble in water.
Percent Purity 99.9995%
Grade UCP-1-M grade, ultra 'F' purity
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Graphite powder, Natural

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.