missing translation for 'onlineSavingsMsg'
Learn More

Graphite powder, natural, briquetting grade, -200 mesh, 99.9995% (metals basis), Thermo Scientific Chemicals

Product Code. 11335077
Change view
Click to view available options
Quantity:
113 g
227 g
454 g
Unit Size:
113g
227g
454g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11335077 454 g 454g
11355077 113 g 113g
11345077 227 g 227g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11335077 Supplier Thermo Scientific Alfa Aesar Supplier No. 040795.TD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Graphite Powder is basically used for casting and polishing purposes, moulding & Carbon addition in steel industries. It is also used for smooth casting iron, copper base alloys and aluminum base alloy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-42-5
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Color Black
Mesh Size −200 Mesh
Odor Odorless
Quantity 454 g
UN Number UN1325
Solubility Information Insoluble in water.
Percent Purity 99.9995%
Grade UCP-1 grade, ultra 'F' purity
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Graphite powder, Natural
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.