missing translation for 'onlineSavingsMsg'
Learn More

Graphite ink for tantalum capacitors, Thermo Scientific Chemicals

Product Code. 11329847
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329847 1 kg 1kg
11339847 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329847 Supplier Thermo Scientific Alfa Aesar Supplier No. 042193.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A graphite ink for use as an undercoat for silver ink in the production of tantalum capacitors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A graphite ink for use as an undercoat for silver ink in the production of tantalum capacitors
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00144065

Specifications

Color Black
Odor Ammonia-like
Quantity 1 kg
Physical Form Paste
Chemical Name or Material Graphite ink for tantalum capacitors

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.