missing translation for 'onlineSavingsMsg'
Learn More

Graphite foil, 0.4mm (0.015in) thick, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog Number 11360228
Click to view available options
Quantity:
15 x 15 cm
15 x 60 cm
15 x 250 cm
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7782-42-5
C
12.01
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
OKTJSMMVPCPJKN-UHFFFAOYSA-N
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
5462310
CHEBI:27594
carbon
[C]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7782-42-5
C
12.01
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
OKTJSMMVPCPJKN-UHFFFAOYSA-N
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
5462310
CHEBI:27594
carbon
[C]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 99.8%, metal basis
Quantity 15 x 15 cm
Physical Form Foil
Chemical Name or Material Graphite

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'