missing translation for 'onlineSavingsMsg'
Learn More

Graphene nanoplatelets aggregates, sub-micronparticles, S.A. 500 m^2/g, Thermo Scientific Chemicals

Product Code. 15427405
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-42-5
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Color Black
Quantity 25 g
Physical Form Nanoplatelets
Chemical Name or Material Graphene nanoplatelets aggregates, sub-micronparticles

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'