Learn More
Pro-Lab Diagnostics™ Gram's Differentiator Solution
Use for convenient Gram staining of bacteria cells. Prolab Diagnostics™ Gram's Differentiator Solution is ready-to-use.
Brand: Pro-Lab Diagnostics™ PL7006
Code : 04
Additional Details : Weight : 0.40500kg Transport : UN number : 1993 Chem class : 3 Pack group : II
Description
- Pro-Lab Diagnostics offers an extensive range of ready-to-use and concentrated microbiological staining solutions
- Solutions come in a selection of convenient and economical sizes.
- All stains are manufactured in accordance with all current regulatory requirements
- CE labelled
Specifications
Reagent | |
500mL | |
Liquid | |
Colorless | |
Acetone, Ethanol, Methanol | |
Gram's Differentiator |
Gram's Differentiator | |
(EC) No 1907/2006, Annex II, (EU) No 453/2010, CE-mark | |
For Gram staining | |
56 to 78°C | |
<20°C |
Safety and Handling
- Grams Differentiator
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H332-Harmful if inhaled.
- H319-Causes serious eye irritation.
- H371-May cause damage to organs.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- P501-Dispose of contents/container to .
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : acetone, ethanol, methanol