missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Gram's Differentiator Solution
Use for convenient Gram staining of bacteria cells. Prolab Diagnostics™ Gram's Differentiator Solution is ready-to-use.
£47.65 - £82.50
Specifications
Solution Type | Reagent |
---|---|
Type | Gram's Differentiator |
Certifications/Compliance | (EC) No 1907/2006, Annex II, (EU) No 453/2010, CE-mark |
Format | Liquid |
For Use With (Application) | For Gram staining |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12996287
|
Pro-Lab Diagnostics™
PL7007 |
1L |
£82.50
1 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12986287
|
Pro-Lab Diagnostics™
PL7006 |
500mL |
£47.65
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Pro-Lab Diagnostics offers an extensive range of ready-to-use and concentrated microbiological staining solutions
- Solutions come in a selection of convenient and economical sizes.
- All stains are manufactured in accordance with all current regulatory requirements
- CE labelled
Specifications
Reagent | |
(EC) No 1907/2006, Annex II, (EU) No 453/2010, CE-mark | |
For Gram staining | |
56 to 78°C | |
<20°C |
Gram's Differentiator | |
Liquid | |
Colorless | |
Acetone, Ethanol, Methanol | |
Gram's Differentiator |