missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Gram Decolorizer
Can be used as a replacement reagent for the BD™ Gram Stain Kits. BD™ Gram Decolorizer is used in a 4-step Gram staining process, before counterstaining. Available in 250ml solution bottles for direct use or gallon containers for bulk storage.
£217.00 - £270.00
Specifications
Type | Decolorizer |
---|---|
Certifications/Compliance | CE |
Format | Liquid |
For Use With (Application) | Staining microorganisms from cultures or specimens by the differential Gram method. |
Color | Clear ad colorless |
Product Code | Brand | Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Quantity | Price | Quantity & Availability | ||||
11798193
|
BD
212527 |
Decolorizer | 4 x 250mL |
£217.00
1000mL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11708203
|
BD
212528 |
Decolorizer | 1 gallon / 3.8L |
£270.00
3.80 L |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Gram Staining is used to differentiate gram-positive and gram-negative microorganisms from cultures or specimens. This Gram Decolorizer is an acetone (25%)/isopropanol (75%) solution for rapid decolorization, prior to counterstaining.
- Complete stain kits reduce the mess associated with Gram staining procedures
- Break-resistant solution bottle features a hinged cap that makes regulating the flow easy and helps prevent spots and spills
- Color-coded label allows quick identification of reagent
Order Info
Available in packs of four 250mL bottles or individual one-gallon bottles
Specifications
Decolorizer | |
Liquid | |
Clear ad colorless | |
15° to 30°C |
CE | |
Staining microorganisms from cultures or specimens by the differential Gram method. | |
No drip flip cap | |
Decolorizer |