Learn More
BD Gram Decolorizer
Can be used as a replacement reagent for the BD™ Gram Stain Kits. BD™ Gram Decolorizer is used in a 4-step Gram staining process, before counterstaining. Available in 250ml solution bottles for direct use or gallon containers for bulk storage.
Brand: BD 212528
Additional Details : CAS Number : 67-63-0 Weight : 3.43800kg Transport : UN number : 1993 Chem class : 3 Pack group : II
Description
Gram Staining is used to differentiate gram-positive and gram-negative microorganisms from cultures or specimens. This Gram Decolorizer is an acetone (25%)/isopropanol (75%) solution for rapid decolorization, prior to counterstaining.
- Complete stain kits reduce the mess associated with Gram staining procedures
- Break-resistant solution bottle features a hinged cap that makes regulating the flow easy and helps prevent spots and spills
- Color-coded label allows quick identification of reagent
Specifications
Decolorizer | |
CE | |
Staining microorganisms from cultures or specimens by the differential Gram method. | |
No drip flip cap | |
15° to 30°C | |
Decolorizer |
1 gallon / 3.8L | |
Liquid | |
Clear ad colorless | |
Acetone (25%), Isopropanol (75%) | |
Bulk storage container |
Safety and Handling
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P240-Ground/bond container and receiving equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
- P405-Store locked up.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol, acetone