missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRAIL/RNF128 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
£177.00 - £409.00
Specifications
| Antigen | GRAIL/RNF128 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.5-2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24356810), Immunohistochemistry-Paraffin 5-10 ug/ml |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18150502
|
Novus Biologicals
NBP2-24610SS |
0.025 mg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18779563
|
Novus Biologicals
NBP2-24610 |
0.1 mg |
£409.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRAIL/RNF128 Polyclonal specifically detects GRAIL/RNF128 in Human, Mouse, Rat, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| GRAIL/RNF128 | |
| Western Blot 0.5-2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24356810), Immunohistochemistry-Paraffin 5-10 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| EC 6.3.2, EC 6.3.2.-, FLJ23516, Gene related to anergy in lymphocytes protein, ring finger protein 128GRAILE3 ubiquitin-protein ligase RNF128 | |
| RNF128 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, horse, chimpanzee, monkey and orangutan, 94% homologous in cow, 88% homologous in cat and 83% homologous in opossum, rat and mouse. |
| 1.0 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat, Canine, Equine, Primate | |
| Q8TEB7 | |
| 79589 | |
| A synthetic peptide corresponding to amino acids portion of amino acids 227-269 of human GRAIL was used as immunogen, GenBank no. NP_919445.1. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title