missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRAIL/RNF128 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP2-24610SS
This item is not returnable.
View return policy
Description
GRAIL/RNF128 Polyclonal specifically detects GRAIL/RNF128 in Human, Mouse, Rat, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| GRAIL/RNF128 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| EC 6.3.2, EC 6.3.2.-, FLJ23516, Gene related to anergy in lymphocytes protein, ring finger protein 128GRAILE3 ubiquitin-protein ligase RNF128 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 79589 | |
| Human, Mouse, Rat, Canine, Equine, Primate | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 0.5-2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24356810), Immunohistochemistry-Paraffin 5-10 ug/ml | |
| Q8TEB7 | |
| RNF128 | |
| A synthetic peptide corresponding to amino acids portion of amino acids 227-269 of human GRAIL was used as immunogen, GenBank no. NP_919445.1. | |
| 0.025 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, horse, chimpanzee, monkey and orangutan, 94% homologous in cow, 88% homologous in cat and 83% homologous in opossum, rat and mouse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction