missing translation for 'onlineSavingsMsg'
Learn More
Learn More
De Buyer™ Stainless-Steel Graduated Bucket
Rinse, carry or store substances with the De Buyer™ Stainless-Steel Graduated Bucket. It is useful for all types of tasks inside and outside of the laboratory.
Brand: De Buyer™ 3274.12N
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Stainless steel
- 10L and 15L capacities
- 1 Each
- Rim with lip
Specifications
28 cm | |
33 cm |