missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Grade C 160 Smooth Qualitative Filter Papers
Filters with increased wet strength for the filtration and absorption of liquids.
£6.30 - £36.50
Specifications
Airflow Rate | 25mbar |
---|---|
Format | Disc |
Material | Refined Pulp |
For Use With (Application) | Used for the filtration of cooking and transformer oils, galvanic baths and as base paper for further impregnation with certain reagents |
Grade | C 160 |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
11720486
|
Sartorius™
FT-3-343-293 |
293 mm |
£36.50
Pack of 50 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
11700486
|
Sartorius™
FT-3-343-142 |
125 mm |
£16.90
Pack of 50 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
11770476
|
Sartorius™
FT-3-343-047 |
47 mm |
£6.30
Pack of 50 |
In Stock
Log in to see stock. |
|||||
Description
Filters with increased wet strength for the filtration and absorption of liquids.Specifications
25mbar | |
Refined Pulp | |
C 160 | |
0.3 mm |
Disc | |
Used for the filtration of cooking and transformer oils, galvanic baths and as base paper for further impregnation with certain reagents | |
50/Pk. | |
Qualitative Filter Boards |