missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Grade C 160 Smooth Qualitative Filter Papers

Brand:  Sartorius™ FT-3-343-293

UNSPSC : 41120000

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11720486

  • £21.80 / Pack of 50


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

50/Pk.
293mm
Refined pulp
0.3mm
25mbar
Qualitative Filter Boards
C 160
Used for the filtration of cooking and transformer oils, galvanic baths and as base paper for further impregnation with certain reagents
Filter discs
293mm