missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 926 Sterilization Paper

Used as wrap for medical clothes.

Brand:  Fisherbrand™ 0926F00024

 View more versions of this product

Product Code. 11794576

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Weight: 60g/m2
  • Creped paper

Applications:

  • Used in steam, gas, gamma-ray sterilization
Specifications

Specifications

Autoclavable
Green
Wrapping of medical clothes, surgical instruments and others articles used in steam, gas and gamma-ray sterilization
Sterilization Paper
926
250 Pk.
EN 868-2
75 x 75 cm
75 cm
60 g/m2
Paper
75 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers