missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 7 Cellulose Fiber and Cotton Linter Extraction Thimble

Suitable for all extractors systems.

Brand:  Fisherbrand™

Code : 47

Additional Details : Weight : 0.19350kg

 View more versions of this product

Product Code. 11754043

  • £105.00 / Pack of 25
In Stock
to see stock.
Add to basket

Description

Description

  • 100% High pure cotton linters fibers
  • Neutral and fat free
  • High mechanical strength
  • Excellent retention capacity

Applications:

  • Extraction of sulphur
  • Extraction of the content of free lipids in food
  • Determination of dust and tar in industrial air and gases
  • Lacquer, binder analysis in paint colours
  • Control of emission smokes in industrial chimneys
  • Gravimetric analysis of dust in gases
Specifications

Specifications

25 mm
Extraction of Sulfur in gaseous masses, determination of dust and tar in industrial air and gases
Suitable for all extractors systems
Extraction Thimble
High Purity Cellulose Fibers and Cotton Linters
7
100 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers