missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 604 Surface Protection Paper

Product Code. 11710005 Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W):
50 m x 46 cm
50 m x 6 cm
520 x 420 mm
570 x 460 mm
Packaging Type:
Roll
Sheet
Unit Size:
Each
Pack of 100
This item is not returnable. View return policy

Product Code. 11710005

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed to protect laboratory surfaces against hazardous spills. Fisherbrand™ Grade 604 Surface Protection Paper guards against damage from acids, toxic, corrosive and radioactive fluids and impacts.

  • Available in sheet or roll form
  • Polyethylene backed 125g/m2 absorbent paper quickly soaks up spills and prevents flow through to work surface

Specifications

Product Type Surface Protection Paper
Material Grade 604 Paper
Length (Metric) 50 m
Width (Metric) 0.6 m
Dimensions (L x W) 50 m x 6 cm
Backing Material Polyethylene
Color White
Thickness 0.25mm
Quantity 1 roll
Packaging Type Roll
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.