missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 602 Medium Filtering Special Filter Paper

Product Code. 11714166 Shop All Fisherbrand Products
Change view
Click to view available options
Format:
Folded Sheet
Sheet
Unit Size:
Pack of 25
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format unitSize
11714166 Sheet Pack of 500
11754566 Folded Sheet Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11714166
Only null left
Add to basket
Add to basket

Suitable for all works in laboratories. Fisherbrand™ 602 Medium Filtering Special Filter Paper is used in filtration of various products, pharmaceutical wrapping (pills, powders, dressing bandages and tablets).

  • Color: Vellum grey

Specifications

Format Sheet
For Use With (Application) All works in laboratories - filtration of various products, pharmaceutical wrapping (pills, powders , dressing bandages and tablets)
Grade 602
Quantity 500 Pk.
Dimensions (L x W) 520 x 420mm
Length (Metric) 520 mm
Width (Metric) 420 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.