missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Grade 6 Smooth Surface Folded Filter Qualitative Filter Papers

Used for analytical and technical purposes.

Brand:  Sartorius™ FT-3-312-110

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10275751

  • £11.10 / Pack of 100
Estimated Shipment: 24-05-2024
to see stock.
Add to basket

Description

Description

Used for analytical and technical purposes
Specifications

Specifications

Disc
15μm/sec.
6
0.17 mm
110 mm
Cotton Linters, Refined Pulp
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications
100 Pack
Qualitative Filter Paper
110 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers