missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Grade 6 Smooth Surface Folded Filter Qualitative Filter Papers
Used for analytical and technical purposes.
£8.10 - £301.00
Specifications
Material | Cotton Linters, Refined Pulp |
---|---|
Filtration Speed | 15μm/sec. |
For Use With (Application) | For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications |
Grade | 6 |
Quantity | 100 Pack |
Product Code | Brand | Format | Diameter (Metric) Disc | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Diameter (Metric) Disc | Price | Quantity & Availability | ||||
10275751
|
Sartorius™
FT-3-312-110 |
Disc | 110 mm |
£11.10
Pack of 100 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
||||
10652412
|
Sartorius™
FT-4-312-110 |
Folded | 110 mm |
£37.50
Pack of 100 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
10776662
|
Sartorius™
FT-3-312-125 |
Disc | 125 mm |
£14.50
Pack of 100 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
10758582
|
Sartorius™
FT-4-312-125 |
Folded | 125 mm |
£37.50
Pack of 100 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
10451812
|
Sartorius™
FT-3-312-150 |
Disc | 150 mm |
£18.60
Pack of 100 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
10092341
|
Sartorius™
FT-4-312-150 |
Folded | 150 mm |
£37.50
Pack of 100 |
In Stock
Log in to see stock. |
||||
10113851
|
Sartorius™
FT-3-312-185 |
Disc | 185 mm |
£24.20
Pack of 100 |
In Stock
Log in to see stock. |
||||
10113901
|
Sartorius™
FT-3-312-210 |
Disc | 210 mm |
£24.20
Pack of 100 |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||
10795892
|
Sartorius™
FT-4-312-185 |
Folded | 185 mm |
£37.50
Pack of 100 |
In Stock
Log in to see stock. |
||||
10595321
|
Sartorius™
FT-4-312-210 |
Folded | 210 mm |
£73.40
Pack of 100 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
10796852
|
Sartorius™
FT-3-312-240 |
Disc | 240 mm |
£36.30
Pack of 100 |
In Stock
Log in to see stock. |
||||
10372212
|
Sartorius™
FT-4-312-240 |
Folded | 240 mm |
£61.80
Pack of 100 |
In Stock
Log in to see stock. |
||||
10082261
|
Sartorius™
FT-3-312-270 |
Disc | 270 mm |
£43.10
Pack of 100 |
In Stock
Log in to see stock. |
||||
10002351
|
Sartorius™
FT-4-312-270 |
Folded | 270 mm |
£61.80
Pack of 100 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
10584191
|
Sartorius™
FT-3-312-320 |
Disc | 320 mm |
£60.60
Pack of 100 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
10397161
|
Sartorius™
FT-4-312-320 |
Folded | 320 mm |
£83.60
Pack of 100 |
In Stock
Log in to see stock. |
||||
10604522
|
Sartorius™
FT-4-312-385 |
Folded | 385 mm |
£301.00
Pack of 100 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
10327551
|
Sartorius™
FT-3-312-385 |
Disc | 385 mm |
£84.90
Pack of 100 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
10255751
|
Sartorius™
FT-3-312-055 |
Disc | 55 mm |
£8.10
Pack of 100 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
10491422
|
Sartorius™
FT-3-312-070 |
Disc | 70 mm |
£8.10
Pack of 100 |
In Stock
Log in to see stock. |
||||
10672582
|
Sartorius™
FT-3-312-090 |
Disc | 90 mm |
£10.20
Pack of 100 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
10748582
|
Sartorius™
FT-4-312-090 |
Folded | 90 mm |
£140.00
Pack of 100 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
Description
Used for analytical and technical purposesSpecifications
Cotton Linters, Refined Pulp | |
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications | |
100 Pack | |
Qualitative Filter Paper |
15μm/sec. | |
6 | |
0.17 mm |