missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Grade 4b, Smooth Paper for Qualitative and Technical Analyses, Sheets

Sartorius Stedim Biotech™ Grade 4b, Smooth Paper for Qualitative and Technical Analyses, Sheets are suitable for analytical and technical purposes and feature medium fast filtering.

Brand:  Sartorius Stedim Biotech™ FT-2-309-210297

Additional Details : Weight : 0.46800kg

 View more versions of this product

Product Code. 15982750

  • £29.35 / Pack of 100

Estimated Shipment: 25-07-2022
to see stock.
Add to basket

Description

Description

  • Made of cotton linters and refined pulp with > 95% alpha-cellulose
  • Medium fast filtering
  • Non-sterile and bulk packed
Specifications

Specifications

Sheet
Cotton Linters and Refined Pulp (>95% alpha-cellulose)
210 x 297 mm
4 b
100/Pk.
8 to 12μm
Qualitative Filter Paper
0.15 μm
75 g/m2
Videos
SDS
Documents
Certificates
Special Offers

Special Offers