missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Grade 292 Qualitative Papers, Sheets
Sartorius Stedim Biotech™ Grade 292 Qualitative Papers, Sheets are especially suitable for qualitative analyses that require a low ash content.
Brand: Sartorius Stedim Biotech™ FT-2-205-120120
Additional Details : Weight : 0.12100kg
Description
- Pure cotton linters or cotton linters with refined pulp
- No additives, such as wet-strengthening agents
Specifications
87 g/m2 | |
Cotton Linters (Low-nitrogen and Nitrates; <0.06% Ash) | |
45 sec. | |
100/Pk. | |
120 mm |
Sheet | |
5 to 8 μm | |
292 | |
0.18 mm | |
120 mm |