missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 1100 Cellulose Technical Filter Paper

Product Code. 11802691 Shop All Fisherbrand Products
Change view
Click to view available options
Format:
Disc
Diameter (Metric):
70 mm
190 mm
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Diameter (Metric) unitSize
11802691 Disc 190 mm Pack of 100
11789945 Disc 70 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11802691

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For filtration of coarse particles and gelatinous precipitates.

  • Creped thin paper
  • Wet strengthened
  • For filtering viscous liquids as arabic gum, gelatine, emulsions like agar-agar, alginate

Specifications

Format Disc
Material Cellulose
Particle Retention 50 to 60 μm
Pore Size 50 to 60 μm
For Use With (Application) Viscous liquids as Arabic gum, Gelatin, emulsions like Agar-agar, Alginate, protective Paper for filter presses
Grade 1100
Quantity 100 Pk.
Thickness (Metric) 0.2 mm
Diameter (Metric) 190 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.