missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gr-1 (Ly-6G/Ly-6C) Rat, anti-Mouse, Alexa Fluor 488, Clone: RB6-8C5, STEMCELL Technologies
Rat Monoclonal Antibody
£168.75 - £386.10
Specifications
| Antigen | Gr-1 (Ly-6G/Ly-6C) |
|---|---|
| Clone | RB6-8C5 |
| Applications | Immunoprecipitation, Western Blot, Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17128601
|
Stemcell Technologies
60028AD.1 |
25 μg |
£168.75
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17118601
|
Stemcell Technologies
60028AD |
100 μg |
£386.10
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The RB6-8C5 antibody reacts with the structurally related murine Ly-6G (21 - 25 kDa) and Ly-6C (14 - 16 kDa) GPI-anchored proteins, which together comprise the granulocyte receptor-1 antigen (Gr-1)
- Gr-1 is expressed on monocytes, neutrophils, subsets of macrophages, plasmacytoid dendritic cells, and T cells
- Monocytes in the bone marrow transiently express Gr-1 during development and the expression level is strongly correlated with granulocyte differentiation and maturation
- In the periphery Gr-1 is found predominantly on neutrophils and is a useful marker for these cells
- Whereas the RB6-8C5 binds to both Ly-6G and Ly-6C, another commonly used anti-Gr-1 antibody, clone 1A8, binds specifically to Ly-6G
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Neutrophil Enrichment Kit (Catalog #19762)
Specifications
| Gr-1 (Ly-6G/Ly-6C) | |
| Immunoprecipitation, Western Blot, Flow Cytometry, Immunocytochemistry | |
| Alexa Fluor 488 | |
| Mouse | |
| Primary |
| RB6-8C5 | |
| Monoclonal | |
| Rat | |
| IgG2b κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title