Learn More
Gr-1 (Ly-6G/Ly-6C) Rat, anti-Mouse, Alexa Fluor 488, Clone: RB6-8C5, STEMCELL Technologies
Rat Monoclonal Antibody
Brand:  Stemcell Technologies 60028AD.1
Encompass_Preferred
 View more versions of this product
Product Code. 17128601
  • £168.75 / 25µg
Description
  • The RB6-8C5 antibody reacts with the structurally related murine Ly-6G (21 - 25 kDa) and Ly-6C (14 - 16 kDa) GPI-anchored proteins, which together comprise the granulocyte receptor-1 antigen (Gr-1)
  • Gr-1 is expressed on monocytes, neutrophils, subsets of macrophages, plasmacytoid dendritic cells, and T cells
  • Monocytes in the bone marrow transiently express Gr-1 during development and the expression level is strongly correlated with granulocyte differentiation and maturation
  • In the periphery Gr-1 is found predominantly on neutrophils and is a useful marker for these cells
  • Whereas the RB6-8C5 binds to both Ly-6G and Ly-6C, another commonly used anti-Gr-1 antibody, clone 1A8, binds specifically to Ly-6G
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Neutrophil Enrichment Kit (Catalog #19762)
Specifications
Specifications
Gr-1 (Ly-6G/Ly-6C)
Monoclonal
Alexa Fluor 488
25 μg
Mouse
Granulocytes and Subsets, Monocytes, Myeloid Cells
Immunoprecipitation, Western Blot, Flow Cytometry, Immunocytochemistry
RB6-8C5
Rat
Primary
IgG2b κ