missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gr-1/Ly-6G Antibody (AP-MAB0866) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£147.00
Specifications
| Antigen | Ly-6G/Ly-6C |
|---|---|
| Clone | AP-MAB0866 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, In vivo assay, CyTOF-ready |
| Applications | Flow Cytometry |
Description
Gr-1/Ly-6G Monoclonal specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, In vivo assay, CyTOF-ready.Specifications
| Ly-6G/Ly-6C | |
| 1 mg/mL | |
| Flow Cytometry | |
| Unconjugated | |
| Rat | |
| Innate Immunity | |
| PBS with No Preservative | |
| LY6G6D | |
| IgG2 | |
| Protein A or G purified |
| AP-MAB0866 | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, In vivo assay, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| 546644 | |
| Mouse spontaneous T lymphoma line TK1 | |
| Primary | |
| Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title