missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gr-1/Ly-6G Antibody (AP-MAB0866) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-06691-0.1mg
This item is not returnable.
View return policy
Description
Gr-1/Ly-6G Monoclonal specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, In vivo assay, CyTOF-ready.
Specifications
| Ly-6G/Ly-6C | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, In vivo assay, CyTOF-ready | |
| LY6G6D | |
| Mouse spontaneous T lymphoma line TK1 | |
| 0.1 mg | |
| Innate Immunity | |
| 546644 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles. | |
| IgG2 |
| Flow Cytometry | |
| AP-MAB0866 | |
| Unconjugated | |
| PBS with No Preservative | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction