missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPRC5D Antibody (3D4), Novus Biologicals™
Human Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | GPRC5D |
|---|---|
| Clone | 3D4 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043038
|
Novus Biologicals
NBP3-26547-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043593
|
Novus Biologicals
NBP3-26547-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPRC5D Monoclonal antibody specifically detects GPRC5D in Human samples. It is validated for ELISASpecifications
| GPRC5D | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| G protein-coupled receptor, family C, group 5, member D, G-protein coupled receptor family C group 5 member D, MGC129713, MGC129714, orphan G-protein coupled receptor | |
| Recombinant Human GPRC5D protein [UniProt Q9NZD1] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 3D4 | |
| ELISA | |
| Unconjugated | |
| Human | |
| GPCR | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| 55507 | |
| IgG4 | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title