missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPRC5D Antibody (3D4), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-26547-50ul
This item is not returnable.
View return policy
Description
GPRC5D Monoclonal antibody specifically detects GPRC5D in Human samples. It is validated for ELISA
Specifications
| GPRC5D | |
| Monoclonal | |
| Unconjugated | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| Human | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 3D4 | |
| ELISA | |
| G protein-coupled receptor, family C, group 5, member D, G-protein coupled receptor family C group 5 member D, MGC129713, MGC129714, orphan G-protein coupled receptor | |
| Recombinant Human GPRC5D protein [UniProt Q9NZD1] | |
| 50 μL | |
| GPCR | |
| 55507 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction