missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPRC5B Recombinant Rabbit Monoclonal Antibody (23GB5710), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | GPRC5B |
|---|---|
| Clone | 23GB5710 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089019
![]() |
Invitrogen™
MA553104 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089669
|
Invitrogen™
MA553105 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPRC5B Recombinant Monoclonal antibody specifically detects GPRC5B in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| GPRC5B | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q923Z0, Q9NZH0 | |
| 293546, 51704, 64297 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5710 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A-69G12.1; G protein-coupled receptor class C group 5 member B; G protein-coupled receptor, class C, group 5, member B; G protein-coupled receptor, family C, group 1, member B; GPRC5B; G-protein coupled receptor family C group 5 member B; RAIG2; RAIG-2; retinoic acid responsive gene protein; retinoic acid-induced gene 2 protein | |
| A synthesized peptide derived from human GPRC5B (350-403AA). | |
| Primary | |
| -20°C | |
| GPRC5B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title