missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPRC5B Recombinant Rabbit Monoclonal Antibody (23GB5710), Invitrogen™
GREENER_CHOICE

Product Code. 30089019
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30089019 100 μL 100µL
30089669 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30089019 Supplier Invitrogen™ Supplier No. MA553104

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

This gene encodes a member of the type 3 G protein-coupled receptor family. Members of this superfamily are characterized by a signature 7-transmembrane domain motif. The encoded protein may modulate insulin secretion and increased protein expression is associated with type 2 diabetes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPRC5B
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5710
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GPRC5B
Gene Accession No. Q923Z0, Q9NZH0
Gene Alias A-69G12.1; G protein-coupled receptor class C group 5 member B; G protein-coupled receptor, class C, group 5, member B; G protein-coupled receptor, family C, group 1, member B; GPRC5B; G-protein coupled receptor family C group 5 member B; RAIG2; RAIG-2; retinoic acid responsive gene protein; retinoic acid-induced gene 2 protein
Gene Symbols GPRC5B
Host Species Rabbit
Immunogen A synthesized peptide derived from human GPRC5B (350-403AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293546, 51704, 64297
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.