missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR35 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
£153.00 - £408.00
Specifications
| Antigen | GPR35 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.5-1.0 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18010014
|
Novus Biologicals
NBP2-24640SS |
0.025 mg |
£153.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18779393
|
Novus Biologicals
NBP2-24640 |
0.1 mg |
£408.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPR35 Polyclonal specifically detects GPR35 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| GPR35 | |
| Western Blot 0.5-1.0 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| G protein-coupled receptor 35, G-protein coupled receptor 35, KYNA receptor, Kynurenic acid receptor | |
| GPR35 | |
| IgG | |
| Immunogen affinity purified |
| 1.0 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| PBS with 0.05% Sodium Azide | |
| 2859 | |
| A portion of amino acids 275-309 of human G Protein-Coupled Receptor GPR35 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title