missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR35 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NBP2-24640SS
This item is not returnable.
View return policy
Description
GPR35 Polyclonal specifically detects GPR35 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| GPR35 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| GPR35 | |
| A portion of amino acids 275-309 of human G Protein-Coupled Receptor GPR35 was used as the immunogen. | |
| 0.025 mg | |
| GPCR | |
| 2859 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 0.5-1.0 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin | |
| G protein-coupled receptor 35, G-protein coupled receptor 35, KYNA receptor, Kynurenic acid receptor | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction