missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR19 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24481-1-AP-150UL
This item is not returnable.
View return policy
Description
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1, 000 members and is the largest known class of molecular targets with proven therapeutic value. GPR19 is an orphan receptor found primarily in the central nervous system. GPR19 was also found in ovary and testis and in human embryonic stem cells. GPR19 has been postulated to play a role in cancer development and increased levels of GPR19 mRNA were found in metastatic melanoma, small cell lung carcinoma and pancreas islet cell carcinoma.Specifications
| GPR19 | |
| Polyclonal | |
| Unconjugated | |
| GPR19 | |
| G protein coupled receptor 19, GPR NGA, GPR19 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14760, 2842 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15760, Q61121 | |
| GPR19 | |
| GPR19 Fusion Protein Ag21619 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction