missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR155 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | GPR155 |
|---|---|
| Concentration | 0.4 mg/mL |
| Applications | Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16870494
|
Proteintech
12659-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16860494
|
Proteintech
12659-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPR155, an integral membrane protein related to G-protein coupled receptors, has been shown to have specific roles in Huntington disease and autism spectrum disorders.Specifications
| GPR155 | |
| Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| Q7Z3F1 | |
| 151556, 68526 | |
| GPR155 Fusion Protein Ag3328 | |
| Primary | |
| -20°C | |
| GPR155 |
| 0.4 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| DEP.7, DEPDC3, G protein coupled receptor 155, GPR155, PGR22 | |
| Gpr155 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title