missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR155 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12659-1-AP-150UL
This item is not returnable.
View return policy
Description
GPR155, an integral membrane protein related to G-protein coupled receptors, has been shown to have specific roles in Huntington disease and autism spectrum disorders.Specifications
| GPR155 | |
| Polyclonal | |
| Unconjugated | |
| GPR155 | |
| DEP.7, DEPDC3, G protein coupled receptor 155, GPR155, PGR22 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 151556, 68526 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q7Z3F1 | |
| Gpr155 | |
| GPR155 Fusion Protein Ag3328 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction