missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR154 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-91966-25ul
This item is not returnable.
View return policy
Description
GPR154 Polyclonal specifically detects GPR154 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GPR154 | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Q6W5P4 | |
| NPSR1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DILDNFNLLPDTQERFYASVIIQNLPALNSAINPLIYCVFSSSISFPCRERRSQDSRMTFRERTERHEMQILSKP | |
| 25 μL | |
| GPCR | |
| 387129 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| G protein-coupled receptor 154, G protein-coupled receptor for asthma susceptibility, GPR154NPSR, GPRAASRT2, G-protein coupled receptor 154, G-protein coupled receptor for asthma susceptibility, G-protein coupled receptor PGR14, neuropeptide S receptor, neuropeptide S receptor 1, PGR14VRR1, vasopressin receptor-related receptor 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction