missing translation for 'onlineSavingsMsg'
Learn More

Poeppelmann™ GPN 910 Series Black LDPE Flat Design Sealing Cover

Product Code. 10246984
Change view
Click to view available options
Size (Metric):
O.D. x H: 62 x 6 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size (Metric) unitSize
10246984 O.D. x H: 62 x 6 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10246984 Supplier Poeppelmann™ Supplier No. 91007150000
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color Black
For Use With (Application) Ideal for closing apertures in cover panels
Length (Metric) 6 mm
Quantity 100 Pack
Size (Metric) O.D. x H: 62 x 6 mm
Diameter (Metric) Outside 62 mm
Material LDPE
Type Sealing Cover
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.