missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GP6 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£161.00 - £333.00
Specifications
| Antigen | GP6 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17980283
|
Invitrogen™
PA5145258 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17909082
![]() |
Invitrogen™
PA5145257 |
200 μL |
£333.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glycoprotein VI (GP6) is a 58-kD platelet membrane glycoprotein that plays a crucial role in the collagen-induced activation and aggregation of platelets. Upon injury to the vessel wall and subsequent damage to the endothelial lining, exposure of the subendothelial matrix to blood flow results in deposition of platelets. Collagen fibers are the most thrombogenic macromolecular components of the extracellular matrix, with collagen types I, III, and VI being the major forms found in blood vessels. Platelet interaction with collagen occurs as a 2-step procedure: (1) the initial adhesion to collagen is followed by (2) an activation step leading to platelet secretion, recruitment of additional platelets, and aggregation. In physiologic conditions, the resulting platelet plug is the initial hemostatic event limiting blood loss. However, exposure of collagen after rupture of atherosclerotic plaques is a major stimulus of thrombus formation associated with myocardial infarction or stroke.Specifications
| GP6 | |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Pig | |
| Q9HCN6 | |
| 100517829, 51206 | |
| Recombinant Human GP6 protein, Tyr86-Thr231 (Accession #Q9HCN6). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| 9830166G18Rik; BDPLT11; Glycoprotein 5; glycoprotein 6; glycoprotein 6 (platelet); glycoprotein VI (platelet); glycoprotein VI platelet; Gm469; GP6; GPIV; GPVI; platelet collagen receptor; platelet glycoprotein VI | |
| GP6 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| GP6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title