missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GP2 Antibody (GP2/1712), Novus Biologicals™
Mouse Monoclonal Antibody
£222.00 - £429.00
Specifications
| Antigen | GP2 |
|---|---|
| Clone | GP2/1712 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18211980
|
Novus Biologicals
NBP2-53322-20UG_TEST |
20 μg |
£222.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18003264
|
Novus Biologicals
NBP2-53322-100UG |
100 μg |
£429.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GP2 Monoclonal specifically detects GP2 in Human, Mouse (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| GP2 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse (Negative) | |
| DKFZp779K0533, glycoprotein 2 (zymogen granule membrane), pancreatic secretory granule membrane major glycoprotein GP2, pancreatic zymogen granule membrane associated protein GP2, Pancreatic zymogen granule membrane protein GP-2, ZAP75 | |
| GP2 | |
| IgG2b λ | |
| Protein A or G purified | |
| 59 kDa |
| GP2/1712 | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 2813 | |
| Recombinant human GP2 protein fragment (aa35-179) (exact sequence is proprietary) | |
| Primary | |
| Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title