missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GP2 Antibody (GP2/1712), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53322-100UG
This item is not returnable.
View return policy
Description
GP2 Monoclonal specifically detects GP2 in Human, Mouse (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| GP2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml | |
| DKFZp779K0533, glycoprotein 2 (zymogen granule membrane), pancreatic secretory granule membrane major glycoprotein GP2, pancreatic zymogen granule membrane associated protein GP2, Pancreatic zymogen granule membrane protein GP-2, ZAP75 | |
| Mouse | |
| 59 kDa | |
| 100 μg | |
| Primary | |
| Human, Mouse (Negative) | |
| Purified |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| GP2/1712 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| GP2 | |
| Recombinant human GP2 protein fragment (aa35-179) (exact sequence is proprietary) | |
| Protein A or G purified | |
| RUO | |
| 2813 | |
| Store at 4C. | |
| IgG2b λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction